Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lats2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015771.2 | NP_056586.2 | 2 - 3 | 773 | 76 | |
GenBank mRNA | AB023958.1 | - | 1 - 2 | 470 | 76 | |
AK029242.1 | - | 754 | 76 | |||
AK029870.1 | - | 2 - 3 | 754 | 76 | ||
AK029966.1 | - | 2 - 3 | 733 | 76 | ||
AK161243.1 | - | 1 - 2 | 195 | 76 | ||
AK163607.1 | - | 2 - 3 | 756 | 76 | ||
AK164789.1 | - | 2 - 3 | 725 | 76 | ||
AK214107.1 | - | 40 | 76 | |||
BC024819.2 | - | 2 - 3 | 764 | 76 | ||
BC040025.1 | - | 1 - 2 | 479 | 76 | ||
BC053028.1 | - | 2 - 3 | 727 | 6409832 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |