Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016712.3 | NP_057921.1 | 5 - 6 | 592 | 65 | |
GenBank mRNA | AF177174.1 | - | 4 - 5 | 511 | 65 | |
AK144408.1 | - | 5 - 6 | 593 | 65 | ||
AK144535.1 | - | 5 - 6 | 593 | 65 | ||
BC068020.1 | - | 4 - 5 | 528 | 30291625 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |