Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016792.4 | NP_058072.2 | 5 - 6 | 747 | 121 | |
GenBank mRNA | AF052660.1 | - | 5 - 6 | 576 | 121 | |
AK029807.1 | - | 5 - 6 | 748 | 121 | ||
AK051698.1 | - | 5 - 6 | 722 | 121 | ||
AK144211.1 | - | 5 - 6 | 704 | 121 | ||
AK150326.1 | - | 5 - 6 | 710 | 121 | ||
AK150887.1 | - | 5 - 6 | 716 | 121 | ||
AK150983.1 | - | 5 - 6 | 716 | 121 | ||
AK152101.1 | - | 5 - 6 | 716 | 121 | ||
AK167954.1 | - | 5 - 6 | 718 | 121 | ||
AK168712.1 | - | 5 - 6 | 711 | 121 | ||
BC061123.1 | - | 5 - 6 | 774 | 6596299 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |