Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016852.2 | NP_058548.1 | 5 - 6 | 572 | 73 | |
GenBank mRNA | AK004824.1 | - | 5 - 6 | 574 | 73 | |
AK043037.1 | - | 5 - 6 | 546 | 73 | ||
AK146572.1 | - | 5 - 6 | 574 | 73 | ||
AK150018.1 | - | 5 - 6 | 547 | 73 | ||
AK151368.1 | - | 5 - 6 | 564 | 73 | ||
AK162810.1 | - | 5 - 6 | 547 | 73 | ||
AK170317.1 | - | 5 - 6 | 573 | 73 | ||
AK170474.1 | - | 5 - 6 | 575 | 73 | ||
AK208273.1 | - | 1 - 2 | 64 | 73 | ||
BC031908.1 | - | 5 - 6 | 535 | 5365118 | 73 | |
BC055058.1 | - | 5 - 6 | 592 | 6529458 | 73 | |
U40826.1 | - | 5 - 6 | 572 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |