Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163522.1 | NP_001156994.1 | 3 - 4 | 434 | 146 | |
NM_016885.2 | NP_058581.2 | 3 - 4 | 434 | 146 | ||
GenBank mRNA | AF060883.1 | - | 3 - 4 | 369 | 146 | |
AK002616.1 | - | 3 - 4 | 416 | 146 | ||
AK164422.1 | - | 355 | 146 | |||
BC003706.1 | - | 3 - 4 | 395 | 3485788 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |