Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159299.2 | NP_001152771.1 | 4 - 5 | 571 | 61 | |
NM_001289632.1 | NP_001276561.1 | 4 - 5 | 571 | 61 | ||
NM_001289633.1 | NP_001276562.1 | 4 - 5 | 571 | 61 | ||
NM_018746.4 | NP_061216.2 | 4 - 5 | 571 | 61 | ||
GenBank mRNA | AF023919.1 | - | 4 - 5 | 534 | 61 | |
AK004862.1 | - | 4 - 5 | 546 | 61 | ||
AK004893.1 | - | 4 - 5 | 546 | 61 | ||
AK050016.1 | - | 4 - 5 | 549 | 61 | ||
AK050270.1 | - | 4 - 5 | 546 | 61 | ||
BC016500.1 | - | 4 - 5 | 534 | 4211735 | 61 | |
BC021459.1 | - | 4 - 5 | 536 | 61 | ||
BC092258.1 | - | 4 - 5 | 523 | 5125112 | 61 | |
BC094457.1 | - | 4 - 5 | 540 | 5136326 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |