Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136071.2 | NP_001129543.1 | 4 - 5 | 563 | 108 | |
NM_001271510.1 | NP_001258439.1 | 4 - 5 | 563 | 108 | ||
NM_019391.3 | NP_062264.1 | 4 - 5 | 930 | 108 | ||
GenBank mRNA | BC044736.1 | - | 4 - 5 | 518 | 108 | |
D49691.1 | - | 4 - 5 | 567 | 108 | ||
M89956.1 | - | 4 - 5 | 505 | 108 | ||
M90316.1 | - | 4 - 5 | 929 | 108 | ||
S74179.1 | - | 4 - 5 | 476 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |