Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019457.2 | NP_062330.1 | 4 - 5 | 556 | 81 | |
GenBank mRNA | AF092208.1 | - | 4 - 5 | 501 | 81 | |
AK016236.1 | - | 4 - 5 | 545 | 81 | ||
AK077124.1 | - | 4 - 5 | 540 | 81 | ||
BC046277.1 | - | 4 - 5 | 507 | 6310345 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |