Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019497.2 | NP_062370.2 | 12 - 13 | 1837 | 68 | |
GenBank mRNA | AF040745.1 | - | 12 - 13 | 1263 | 68 | |
AK132685.1 | - | 12 - 13 | 1838 | 68 | ||
BC150691.1 | - | 12 - 13 | 1361 | 9087602 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |