Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcerg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039474.1 | NP_001034563.1 | 18 - 19 | 2780 | 72 | |
NM_001289526.1 | NP_001276455.1 | 18 - 19 | 2768 | 72 | ||
NR_110344.1 | 18 - 19 | 2780 | 72 | |||
GenBank mRNA | AB023485.1 | - | 18 - 19 | 2744 | 72 | |
AK076993.1 | - | 18 - 19 | 2761 | 72 | ||
AK082750.1 | - | 2 - 3 | 264 | 72 | ||
AK168434.1 | - | 18 - 19 | 2758 | 72 | ||
BC039185.1 | - | 17 - 18 | 2670 | 3982736 | 72 | |
BC040284.1 | - | 17 - 18 | 2680 | 4460736 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |