Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pias1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019663.3 | NP_062637.2 | 4 - 5 | 604 | 75 | |
GenBank mRNA | AF077950.1 | - | 4 - 5 | 596 | 75 | |
AK075708.1 | - | 4 - 5 | 605 | 75 | ||
AK153871.1 | - | 4 - 5 | 609 | 75 | ||
BC051417.1 | - | 4 - 5 | 601 | 6515027 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |