Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161714.1 | NP_001155186.1 | 12 - 13 | 2034 | 74 | |
NM_001161715.1 | NP_001155187.1 | 12 - 13 | 2133 | 74 | ||
NM_019984.3 | NP_064368.3 | 12 - 13 | 2082 | 74 | ||
GenBank mRNA | AF186373.1 | - | 12 - 13 | 2025 | 74 | |
BC026422.1 | - | 12 - 13 | 2082 | 4221991 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |