Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021322.2 | NP_067297.2 | 6 - 7 | 871 | 69 | |
GenBank mRNA | AJ271892.1 | - | 6 - 7 | 641 | 69 | |
AJ271893.1 | - | 6 - 7 | 641 | 69 | ||
AK035125.1 | - | 5 - 6 | 495 | 69 | ||
AK144821.1 | - | 6 - 7 | 676 | 69 | ||
AK152271.1 | - | 6 - 7 | 663 | 69 | ||
AK153161.1 | - | 6 - 7 | 663 | 69 | ||
AK164160.1 | - | 6 - 7 | 584 | 69 | ||
BC039272.1 | - | 6 - 7 | 671 | 5027600 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |