Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033621.3 | NP_001028793.1 | 2 - 3 | 621 | 90 | |
GenBank mRNA | AF230979.2 | - | 2 - 3 | 620 | 90 | |
AK017447.1 | - | 2 - 3 | 622 | 90 | ||
BC016214.1 | - | 2 - 3 | 599 | 4482382 | 90 | |
BC064052.1 | - | 2 - 3 | 594 | 1366418 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |