Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033621.3 | NP_001028793.1 | 6 - 7 | 1077 | 96 | |
GenBank mRNA | AF230979.2 | - | 6 - 7 | 1076 | 96 | |
AK017447.1 | - | 6 - 7 | 1078 | 96 | ||
BC016214.1 | - | 6 - 7 | 1055 | 4482382 | 96 | |
BC064052.1 | - | 6 - 7 | 1050 | 1366418 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |