Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpa33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021610.1 | NP_067623.1 | 1 - 2 | 144 | 94 | |
GenBank mRNA | AF247659.1 | - | 1 - 2 | 144 | 94 | |
AK008784.1 | - | 1 - 2 | 176 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons |