Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021879.2 | NP_068679.1 | 6 - 7 | 762 | 97 | |
GenBank mRNA | AK051897.1 | - | 6 - 7 | 757 | 97 | |
AK087494.1 | - | 7 - 8 | 1022 | 97 | ||
BC119220.1 | - | 6 - 7 | 732 | 8733969 | 97 | |
BC120549.1 | - | 6 - 7 | 732 | 40129472 | 97 | |
M97900.1 | - | 6 - 7 | 762 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |