Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100011587.mm.1 |
NM_001160181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011587.mm.1 |
NM_001160181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011587.mm.1 |
NM_022329.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011587.mm.1 |
NM_022329.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011587.mm.1 |
TC0100001409.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_001160180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_001160180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_001160181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_001160181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_001160182.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_001160182.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_022329.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
NM_022329.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011593.mm.1 |
TC0100001409.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011583.mm.1 |
NM_001160181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011583.mm.1 |
NM_001160181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011583.mm.1 |
NM_022329.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011583.mm.1 |
NM_022329.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011583.mm.1 |
TC0100001409.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_001160180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_001160180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_001160181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_001160181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_001160182.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_001160182.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_022329.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
NM_022329.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011595.mm.1 |
TC0100001409.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_001160180.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_001160180.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_001160181.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_001160181.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_001160182.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_001160182.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_022329.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
NM_022329.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100011592.mm.1 |
TC0100001409.mm.1 |
MTA TC |