Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199118.1 | NP_001186047.1 | 10 - 11 | 1705 | 81 | |
NM_022883.3 | NP_075021.1 | 10 - 11 | 1782 | 81 | ||
GenBank mRNA | AF286724.1 | - | 10 - 11 | 1719 | 81 | |
AK033647.1 | - | 10 - 11 | 1703 | 81 | ||
AK049363.1 | - | 10 - 11 | 1698 | 81 | ||
AK163833.1 | - | 10 - 11 | 1697 | 81 | ||
AK220228.1 | - | 10 - 11 | 1782 | 81 | ||
BC026729.1 | - | 10 - 11 | 1689 | 81 | ||
BC117882.1 | - | 9 - 10 | 1646 | 40096038 | 81 | |
BC117883.1 | - | 9 - 10 | 1646 | 40096041 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |