Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025375.2 | NP_001020546.1 | 7 - 8 | 782 | 72 | |
NM_001025376.2 | NP_001020547.1 | 7 - 8 | 671 | 72 | ||
NM_023191.3 | NP_075680.1 | 7 - 8 | 676 | 72 | ||
GenBank mRNA | AF309554.1 | - | 7 - 8 | 634 | 72 | |
AK010283.1 | - | 7 - 8 | 622 | 72 | ||
AK012342.1 | - | 7 - 8 | 688 | 72 | ||
AK013114.1 | - | 7 - 8 | 640 | 72 | ||
AK077037.1 | - | 716 | 72 | |||
AK142498.1 | - | 7 - 8 | 639 | 72 | ||
AK153862.1 | - | 7 - 8 | 709 | 72 | ||
AK167706.1 | - | 7 - 8 | 709 | 72 | ||
AK205840.1 | - | 1 - 2 | 68 | 72 | ||
BC023026.1 | - | 7 - 8 | 621 | 5374890 | 72 | |
EF011623.1 | - | 7 - 8 | 619 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |