Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085383.1 | NP_001078852.1 | 5 - 6 | 521 | 63 | |
NM_023628.2 | NP_076117.2 | 5 - 6 | 500 | 63 | ||
GenBank mRNA | AJ401160.1 | - | 5 - 6 | 454 | 63 | |
AK003395.1 | - | 5 - 6 | 521 | 63 | ||
AK010128.1 | - | 5 - 6 | 501 | 63 | ||
BC062140.1 | - | 5 - 6 | 511 | 30290956 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |