Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159965.1 | NP_001153437.1 | 11 - 12 | 1126 | 115 | |
NM_001159966.1 | NP_001153438.1 | 11 - 12 | 1024 | 115 | ||
NM_001159967.1 | NP_001153439.1 | 11 - 12 | 1126 | 115 | ||
NM_001159968.1 | NP_001153440.1 | 10 - 11 | 1021 | 115 | ||
NM_023884.4 | NP_076373.3 | 11 - 12 | 1066 | 115 | ||
NR_027640.1 | 12 - 13 | 1231 | 115 | |||
GenBank mRNA | AF312924.1 | - | 10 - 11 | 974 | 115 | |
AK018622.1 | - | 11 - 12 | 1068 | 115 | ||
AK019543.1 | - | 10 - 11 | 1024 | 115 | ||
AK029049.1 | - | 11 - 12 | 1067 | 115 | ||
AK029993.1 | - | 12 - 13 | 1204 | 115 | ||
AK033549.1 | - | 11 - 12 | 1022 | 115 | ||
AK036803.1 | - | 10 - 11 | 998 | 115 | ||
AK161080.1 | - | 11 - 12 | 1093 | 115 | ||
AK165002.1 | - | 11 - 12 | 1018 | 115 | ||
AK169368.1 | - | 11 - 12 | 1044 | 115 | ||
BC043132.1 | - | 11 - 12 | 1031 | 6811441 | 115 | |
BC052663.1 | - | 11 - 12 | 30051209 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |