Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029573.2 | NP_083849.1 | 7 - 8 | 722 | 148 | |
GenBank mRNA | AK003393.1 | - | 4 - 5 | 615 | 148 | |
AK010065.1 | - | 7 - 8 | 746 | 148 | ||
AK032787.1 | - | 7 - 8 | 746 | 148 | ||
AK047951.1 | - | 7 - 8 | 744 | 148 | ||
AK117103.1 | - | 7 - 8 | 758 | 148 | ||
AK146717.1 | - | 4 - 5 | 586 | 148 | ||
AK150618.1 | - | 7 - 8 | 753 | 148 | ||
AK151304.1 | - | 7 - 8 | 747 | 148 | ||
AK152353.1 | - | 7 - 8 | 756 | 148 | ||
AK153459.1 | - | 7 - 8 | 747 | 148 | ||
AK158646.1 | - | 7 - 8 | 900 | 148 | ||
AK159051.1 | - | 7 - 8 | 819 | 148 | ||
AK159666.1 | - | 7 - 8 | 753 | 148 | ||
AK160054.1 | - | 7 - 8 | 753 | 148 | ||
AK164038.1 | - | 8 - 9 | 849 | 149 | ||
AK167822.1 | - | 7 - 8 | 752 | 148 | ||
AK168034.1 | - | 7 - 8 | 744 | 148 | ||
AK168049.1 | - | 7 - 8 | 747 | 148 | ||
AK168149.1 | - | 7 - 8 | 753 | 148 | ||
AK169152.1 | - | 7 - 8 | 720 | 148 | ||
BC034273.1 | - | 7 - 8 | 712 | 4166193 | 148 | |
BC049956.1 | - | 7 - 8 | 722 | 4212315 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |