Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK148247.1 | - | 2 - 3 | 138 | 69 | |
AK171019.1 | - | 2 - 3 | 205 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |