Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053273.2 | NP_444503.2 | 3 - 4 | 487 | 63 | |
GenBank mRNA | AF329682.1 | - | 3 - 4 | 430 | 63 | |
AK152172.1 | - | 3 - 4 | 439 | 63 | ||
AK154835.1 | - | 3 - 4 | 495 | 63 | ||
AK155166.1 | - | 3 - 4 | 488 | 63 | ||
AK156393.1 | - | 3 - 4 | 440 | 63 | ||
AK168408.1 | - | 3 - 4 | 440 | 63 | ||
BC060740.1 | - | 3 - 4 | 430 | 4158813 | 63 | |
BY740853.1 | - | 3 - 4 | 467 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |