Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146323.1 | NP_001139795.1 | 13 - 14 | 2129 | 88 | |
NM_080634.4 | NP_542365.3 | 13 - 14 | 2525 | 88 | ||
GenBank mRNA | AF393747.1 | - | 13 - 14 | 2514 | 88 | |
AF393780.1 | - | 13 - 14 | 2517 | 88 | ||
AK034377.1 | - | 13 - 14 | 2496 | 88 | ||
AK036663.1 | - | 13 - 14 | 2533 | 88 | ||
AK155497.1 | - | 13 - 14 | 2493 | 88 | ||
AK157953.1 | - | 13 - 14 | 2511 | 88 | ||
AK169855.1 | - | 12 - 13 | 2597 | 88 | ||
AK170581.1 | - | 13 - 14 | 2041 | 88 | ||
AK171997.1 | - | 13 - 14 | 2117 | 88 | ||
BC020105.1 | - | 3 - 4 | 624 | 88 | ||
BC038087.1 | - | 3 - 4 | 616 | 88 | ||
BC049350.1 | - | 4 - 5 | 716 | 88 | ||
BC058235.1 | - | 13 - 14 | 2499 | 6307400 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |