Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Recql5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130454.2 | NP_569721.1 | 4 - 5 | 1451 | 103 | |
GenBank mRNA | AB038562.1 | - | 2 - 3 | 348 | 103 | |
AB059999.1 | - | 4 - 5 | 965 | 103 | ||
AK029060.1 | - | 4 - 5 | 1105 | 103 | ||
AK050965.1 | - | 2 - 3 | 180 | 103 | ||
AK083513.1 | - | 4 - 5 | 1447 | 103 | ||
BC023769.1 | - | 3 - 4 | 743 | 103 | ||
BC053086.1 | - | 4 - 5 | 1704 | 103 | ||
BC059000.1 | - | 4 - 5 | 1425 | 103 | ||
BC060070.1 | - | 4 - 5 | 1704 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |