Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdhr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130878.2 | NP_570948.1 | 1 - 2 | 230 | 66 | |
GenBank mRNA | AB093300.1 | - | 1 - 2 | 646 | 66 | |
AF426393.1 | - | 1 - 2 | 117 | 66 | ||
AK143151.1 | - | 1 - 2 | 169 | 66 | ||
BC042454.1 | - | 1 - 2 | 140 | 5369576 | 66 | |
BC058270.1 | - | 1 - 2 | 230 | 6490597 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |