Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133199.2 | NP_573462.2 | 2 - 3 | 391 | 72 | |
GenBank mRNA | AJ278787.1 | - | 2 - 3 | 391 | 72 | |
BC129805.1 | - | 2 - 3 | 389 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |