Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007585.3 | NP_031611.1 | 7 - 8 | 608 | 73 | |
GenBank mRNA | AK088645.1 | - | 7 - 8 | 597 | 73 | |
AK146096.1 | - | 7 - 8 | 596 | 73 | ||
AK150586.1 | - | 7 - 8 | 596 | 73 | ||
AK152561.1 | - | 7 - 8 | 596 | 73 | ||
AK160746.1 | - | 7 - 8 | 596 | 73 | ||
AK164507.1 | - | 7 - 8 | 601 | 73 | ||
AK165888.1 | - | 7 - 8 | 596 | 73 | ||
AK167894.1 | - | 7 - 8 | 595 | 73 | ||
AK168083.1 | - | 7 - 8 | 596 | 73 | ||
BC003327.1 | - | 7 - 8 | 608 | 3156634 | 73 | |
BC004659.1 | - | 3 - 4 | 3497991 | 73 | ||
BC005763.1 | - | 7 - 8 | 582 | 2655513 | 73 | |
D10024.1 | - | 6 - 7 | 541 | 73 | ||
M14044.1 | - | 6 - 7 | 541 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |