Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007608.2 | NP_031634.2 | 4 - 5 | 653 | 57 | |
GenBank mRNA | AI098626.1 | - | 4 - 5 | 569 | 57 | |
AI314492.1 | - | 4 - 5 | 635 | 57 | ||
BC030174.1 | - | 4 - 5 | 631 | 5051543 | 57 | |
X51971.1 | - | 4 - 5 | 642 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |