Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008292.4 | NP_032318.2 | 4 - 5 | 428 | 61 | |
GenBank mRNA | AK004866.1 | - | 4 - 5 | 428 | 61 | |
AK088381.1 | - | 4 - 5 | 403 | 61 | ||
AK147182.1 | - | 4 - 5 | 400 | 61 | ||
AK152532.1 | - | 4 - 5 | 323 | 61 | ||
AK161655.1 | - | 4 - 5 | 323 | 61 | ||
AK166801.1 | - | 4 - 5 | 341 | 61 | ||
AK167060.1 | - | 4 - 5 | 378 | 61 | ||
AK169077.1 | - | 4 - 5 | 330 | 61 | ||
BC022175.1 | - | 4 - 5 | 301 | 4209394 | 61 | |
X89998.1 | - | 4 - 5 | 289 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |