Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198635.1 | NP_001185564.1 | 8 - 9 | 996 | 69 | |
NM_008635.2 | NP_032661.2 | 8 - 9 | 972 | 69 | ||
GenBank mRNA | AB098611.1 | - | 8 - 9 | 878 | 69 | |
AK014575.1 | - | 8 - 9 | 889 | 69 | ||
AK133267.1 | - | 8 - 9 | 1421 | 69 | ||
BC042771.1 | - | 8 - 9 | 897 | 69 | ||
BC044733.1 | - | 8 - 9 | 897 | 69 | ||
BC046555.1 | - | 8 - 9 | 897 | 69 | ||
BC052637.1 | - | 8 - 9 | 932 | 6467153 | 69 | |
Y15197.1 | - | 8 - 9 | 878 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |