Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285489.1 | NP_001272418.1 | 7 - 8 | 630 | 87 | |
NM_001285490.1 | NP_001272419.1 | 7 - 8 | 643 | 87 | ||
NM_008672.3 | NP_032698.1 | 7 - 8 | 630 | 87 | ||
GenBank mRNA | AJ002198.1 | - | 7 - 8 | 600 | 87 | |
AK090112.1 | - | 7 - 8 | 646 | 87 | ||
AK219914.1 | - | 2 - 3 | 196 | 87 | ||
BC089018.1 | - | 7 - 8 | 616 | 87 | ||
BC132039.1 | - | 6 - 7 | 554 | 40130316 | 87 | |
BC138518.1 | - | 6 - 7 | 554 | 8861538 | 87 | |
BC145306.1 | - | 6 - 7 | 554 | 9053860 | 87 | |
BC145308.1 | - | 6 - 7 | 554 | 9053862 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |