Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285489.1 | NP_001272418.1 | 8 - 9 | 700 | 120 | |
NM_001285490.1 | NP_001272419.1 | 8 - 9 | 713 | 120 | ||
NM_008672.3 | NP_032698.1 | 8 - 9 | 700 | 120 | ||
GenBank mRNA | AJ002198.1 | - | 8 - 9 | 670 | 120 | |
AK090112.1 | - | 8 - 9 | 716 | 120 | ||
AK156835.1 | - | 8 - 9 | 673 | 120 | ||
BC089018.1 | - | 8 - 9 | 686 | 120 | ||
BC132039.1 | - | 7 - 8 | 624 | 40130316 | 120 | |
BC138518.1 | - | 7 - 8 | 624 | 8861538 | 120 | |
BC145306.1 | - | 7 - 8 | 624 | 9053860 | 120 | |
BC145308.1 | - | 7 - 8 | 624 | 9053862 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |