Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB219976.1 | - | 12 - 13 | 2951 | 75 | |
AF000582.1 | - | 15 - 16 | 3312 | 75 | ||
AK035719.1 | - | 2 - 3 | 289 | 75 | ||
BC053387.1 | - | 14 - 15 | 3296 | 30054358 | 75 | |
BU703942.1 | - | 4 - 5 | 614 | 75 | ||
CK637131.1 | - | 3 - 4 | 413 | 75 | ||
U39060.1 | - | 15 - 16 | 3361 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |