Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113209.2 | NP_001106680.1 | 6 - 7 | 1938 | 72 | |
NM_001113210.2 | NP_001106681.1 | 6 - 7 | 1938 | 72 | ||
NM_001286127.1 | NP_001273056.1 | 6 - 7 | 1938 | 72 | ||
NM_001286131.1 | NP_001273060.1 | 6 - 7 | 1938 | 72 | ||
NM_008687.6 | NP_032713.3 | 6 - 7 | 1938 | 72 | ||
GenBank mRNA | AK034261.1 | - | 6 - 7 | 1470 | 72 | |
AK083167.1 | - | 6 - 7 | 1283 | 72 | ||
AK084941.1 | - | 6 - 7 | 1543 | 72 | ||
AK087480.1 | - | 6 - 7 | 1470 | 72 | ||
AK145267.1 | - | 5 - 6 | 644 | 72 | ||
AK146480.1 | - | 6 - 7 | 1455 | 72 | ||
AY035852.1 | - | 6 - 7 | 943 | 72 | ||
AY035853.1 | - | 6 - 7 | 943 | 72 | ||
BC014290.1 | - | 6 - 7 | 975 | 4038233 | 72 | |
BC023366.1 | - | 6 - 7 | 1075 | 72 | ||
BC062908.1 | - | 6 - 7 | 1260 | 6815775 | 72 | |
U57634.1 | - | 6 - 7 | 1144 | 72 | ||
Y07685.1 | - | 6 - 7 | 1870 | 72 | ||
Y07686.1 | - | 6 - 7 | 1450 | 72 | ||
Y07687.1 | - | 6 - 7 | 1931 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |