Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008976.2 | NP_033002.2 | 2 - 3 | 708 | 104 | |
GenBank mRNA | AF170902.1 | - | 1 - 2 | 275 | 104 | |
AF170903.1 | - | 1 - 2 | 275 | 104 | ||
AF170904.1 | - | 1 - 2 | 275 | 104 | ||
AK028783.1 | - | 2 - 3 | 672 | 104 | ||
AK037646.1 | - | 2 - 3 | 709 | 104 | ||
AK086524.1 | - | 2 - 3 | 702 | 104 | ||
AK156480.1 | - | 2 - 3 | 454 | 104 | ||
D31842.1 | - | 1 - 2 | 305 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mus musculus 1700 Probe spans exons |