Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpn14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008976.2 | NP_033002.2 | 10 - 11 | 1470 | 137 | |
GenBank mRNA | AF170902.1 | - | 9 - 10 | 1037 | 137 | |
AF170903.1 | - | 9 - 10 | 1037 | 137 | ||
AK028783.1 | - | 10 - 11 | 1434 | 137 | ||
AK037646.1 | - | 10 - 11 | 1471 | 137 | ||
AK156480.1 | - | 10 - 11 | 1216 | 137 | ||
D31842.1 | - | 9 - 10 | 1067 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mus musculus Probe spans exons |