Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198914.1 | NP_001185843.1 | 8 - 9 | 1213 | 105 | |
NM_010848.3 | NP_034978.3 | 8 - 9 | 1213 | 105 | ||
GenBank mRNA | AK088079.1 | - | 8 - 9 | 1134 | 105 | |
AK088598.1 | - | 8 - 9 | 1135 | 105 | ||
AK132250.1 | - | 8 - 9 | 1078 | 105 | ||
AK137794.1 | - | 2 - 3 | 158 | 105 | ||
AK137965.1 | - | 8 - 9 | 1079 | 105 | ||
BC011513.1 | - | 8 - 9 | 1121 | 3672769 | 105 | |
M12848.1 | - | 8 - 9 | 1211 | 105 | ||
X02774.1 | - | 8 - 9 | 983 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |