Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Per1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159367.1 | NP_001152839.1 | 14 - 15 | 1905 | 89 | |
NM_011065.4 | NP_035195.2 | 14 - 15 | 1920 | 89 | ||
GenBank mRNA | AB002108.1 | - | 13 - 14 | 1734 | 89 | |
AF022992.1 | - | 14 - 15 | 1959 | 89 | ||
AK081813.1 | - | 14 - 15 | 1918 | 89 | ||
AK148202.1 | - | 14 - 15 | 1912 | 89 | ||
AK154900.1 | - | 14 - 15 | 1901 | 89 | ||
AK172958.1 | - | 14 - 15 | 1905 | 89 | ||
BC039768.1 | - | 13 - 14 | 1805 | 4163009 | 89 | |
BC091645.1 | - | 14 - 15 | 1907 | 30355259 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |