Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 29 - 30 | 4350 | 71 | |
GenBank mRNA | AF093576.1 | - | 29 - 30 | 4258 | 71 | |
BC150747.1 | - | 29 - 30 | 4215 | 9087658 | 71 | |
U87455.1 | - | 29 - 30 | 4330 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |