Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011465.4 | NP_035595.2 | 37 - 38 | 5464 | 59 | |
GenBank mRNA | AF093576.1 | - | 37 - 38 | 5372 | 59 | |
AK143823.1 | - | 1 - 2 | 131 | 59 | ||
AK161449.1 | - | 8 - 9 | 1105 | 59 | ||
BC150747.1 | - | 37 - 38 | 5329 | 9087658 | 59 | |
M10276.1 | - | 5 - 6 | 601 | 59 | ||
U87455.1 | - | 37 - 38 | 5444 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |