Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011715.2 | NP_035845.1 | 9 - 10 | 1107 | 81 | |
GenBank mRNA | AF020055.1 | - | 9 - 10 | 1107 | 81 | |
AK004644.1 | - | 9 - 10 | 1215 | 81 | ||
AK004858.1 | - | 9 - 10 | 1178 | 81 | ||
AK151146.1 | - | 9 - 10 | 1104 | 81 | ||
AK153338.1 | - | 9 - 10 | 1118 | 81 | ||
AK156300.1 | - | 9 - 10 | 1144 | 81 | ||
AK165225.1 | - | 9 - 10 | 1077 | 81 | ||
AK165323.1 | - | 9 - 10 | 1078 | 81 | ||
AK167241.1 | - | 9 - 10 | 1133 | 81 | ||
AK167275.1 | - | 9 - 10 | 1175 | 81 | ||
AK167813.1 | - | 9 - 10 | 1105 | 81 | ||
AK169835.1 | - | 9 - 10 | 1201 | 81 | ||
AK169893.1 | - | 9 - 10 | 1137 | 81 | ||
AK170379.1 | - | 9 - 10 | 1117 | 81 | ||
BC119358.1 | - | 9 - 10 | 1063 | 8734107 | 81 | |
BC125295.1 | - | 9 - 10 | 1063 | 40129810 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |