Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpxm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018867.5 | NP_061355.3 | 10 - 11 | 1634 | 87 | |
GenBank mRNA | AF017639.1 | - | 10 - 11 | 1635 | 87 | |
AK019509.1 | - | 10 - 11 | 1633 | 87 | ||
BC021444.1 | - | 10 - 11 | 1556 | 5038528 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |