Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019587.2 | NP_062533.2 | 9 - 10 | 2036 | 58 | |
GenBank mRNA | AK122467.1 | - | 8 - 9 | 962 | 58 | |
AK138631.1 | - | 9 - 10 | 1111 | 58 | ||
AK140240.1 | - | 9 - 10 | 2040 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |