Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285891.1 | NP_001272820.1 | 7 - 8 | 1031 | 82 | |
NM_001285892.1 | NP_001272821.1 | 7 - 8 | 1031 | 82 | ||
NM_019748.3 | NP_062722.1 | 7 - 8 | 1031 | 82 | ||
GenBank mRNA | AB024303.1 | - | 7 - 8 | 919 | 82 | |
AK010313.1 | - | 7 - 8 | 908 | 82 | ||
AK011772.1 | - | 7 - 8 | 942 | 82 | ||
AK011783.1 | - | 4 - 5 | 378 | 82 | ||
AK087556.1 | - | 7 - 8 | 922 | 82 | ||
AK090012.1 | - | 7 - 8 | 918 | 82 | ||
AK154139.1 | - | 7 - 8 | 944 | 82 | ||
AK159672.1 | - | 7 - 8 | 933 | 82 | ||
AK162789.1 | - | 7 - 8 | 919 | 82 | ||
BC068164.1 | - | 7 - 8 | 920 | 30359189 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |