Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB028920.1 | - | 2 - 3 | 91 | 86 | |
AK004801.1 | - | 2 - 3 | 97 | 86 | ||
AK089092.1 | - | 2 - 3 | 104 | 86 | ||
BC042602.1 | - | 2 - 3 | 87 | 86 | ||
BC060170.1 | - | 2 - 3 | 82 | 86 | ||
BC060209.1 | - | 2 - 3 | 188 | 86 | ||
BC078448.1 | - | 2 - 3 | 75 | 86 | ||
BC125521.1 | - | 2 - 3 | 87 | 40130036 | 86 | |
BC137746.1 | - | 2 - 3 | 87 | 8860765 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |