Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Piwil2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021308.1 | NP_067283.1 | 12 - 13 | 1651 | 60 | |
GenBank mRNA | AB032605.1 | - | 12 - 13 | 1651 | 60 | |
AF285586.1 | - | 3 - 4 | 380 | 60 | ||
AK030116.1 | - | 12 - 13 | 1595 | 60 | ||
BC138444.1 | - | 12 - 13 | 1639 | 8861464 | 60 | |
BC145717.1 | - | 12 - 13 | 1639 | 40131020 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |