Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163511.1 | NP_001156983.1 | 10 - 11 | 1937 | 102 | |
NM_021343.2 | NP_067318.2 | 10 - 11 | 1934 | 102 | ||
GenBank mRNA | AF049099.1 | - | 10 - 11 | 1916 | 102 | |
AK011111.2 | - | 10 - 11 | 1901 | 102 | ||
AK144998.1 | - | 10 - 11 | 2264 | 102 | ||
BC145302.1 | - | 10 - 11 | 1905 | 9053856 | 102 | |
BC145309.1 | - | 10 - 11 | 1905 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |